missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail
Specifications
Specifications
Absorbance | ≤0.10 (0.1% Solution) at 260nm |
Color | White |
Physical Form | Solid |
Melting Point | 204°C |
pH | 8.5 to 10 |
Quantity | 500 g |
Identification | Pass test |
Packaging | Poly Bottle |
Assay Percent Range | >99% |
IUPAC Name | sodium;dodecyl sulfate |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling



Product Identifier
- Dodecyl sulfate sodium salt
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction