Learn More
Sodium hexafluoroantimonate, 98%, Thermo Scientific Chemicals
£33.90 - £91.40
Chemical Identifiers
CAS | 16925-25-0 |
---|---|
Molecular Formula | F6NaSb |
Molecular Weight (g/mol) | 258.72 |
MDL Number | MFCD00003482 |
InChI Key | HKLMYZVMEYYVBS-UHFFFAOYSA-H |
Synonym | sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt |
PubChem CID | 16689647 |
IUPAC Name | sodium;hexafluoroantimony(1-) |
SMILES | F[Sb-](F)(F)(F)(F)F.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10471713
|
Thermo Scientific Acros
|
25 g | Plastic bottle |
£33.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10485081
|
Thermo Scientific Acros
419541000 |
100 g | Plastic bottle |
£91.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
16925-25-0 | |
258.72 | |
HKLMYZVMEYYVBS-UHFFFAOYSA-H | |
16689647 | |
F[Sb-](F)(F)(F)(F)F.[Na+] |
F6NaSb | |
MFCD00003482 | |
sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt | |
sodium;hexafluoroantimony(1-) |
Specifications
16925-25-0 | |
Fine Crystalline Powder | |
F6NaSb | |
MFCD00003482 | |
Solubility in water: very soluble | |
F[Sb-](F)(F)(F)(F)F.[Na+] | |
258.72 | |
258.72 | |
Sodium hexafluoroantimonate, 98% |
White | |
97.5% min | |
NaSbF6 | |
sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt | |
HKLMYZVMEYYVBS-UHFFFAOYSA-H | |
sodium;hexafluoroantimony(1-) | |
16689647 | |
98% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 240-989-8
missing translation for 'tsca' : TSCA
- Sodium hexafluoroantimonate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only