missing translation for 'onlineSavingsMsg'
Learn More
Sodium hydroxide, for analysis, 50 wt% solution in water, Thermo Scientific Chemicals Product Code.: 10371503

Sodium hydroxide, for analysis, 50 wt% solution in water, Thermo Scientific Chemicals

Product Code. 10371503
500 mL, Plastic bottle
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10371503

Brand: Thermo Scientific Acros 380215000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, causticShow MoreShow Less
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Boiling Point 145.0°C
Melting Point 12.0°C
Linear Formula NaOH
Density 1.5000g/mL
Merck Index 15, 8761
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone and ether
Quantity 500 mL
Formula Weight 40
Percent Purity 49 to 51%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.5
Physical Form Viscous Solution
Chemical Name or Material Sodium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465894 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number380210100,380210025,380215000
Lot NumberA0468043 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number380210100,380210025,380215000
Lot NumberA0467194 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number380210100,380210025,380215000
Lot NumberA0462485 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number380210100,380210025,380215000
Lot NumberA0462021 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number380210100,380210025,380215000
compliance-icons
Product Identifier
  • Sodium hydroxide, 50 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydroxide, for analysis, 50 wt% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.