missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N)
£18.79 - £199.33
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15634910
|
Honeywell-Fluka
35263-1L |
1 L | Plastic Bottle |
£18.79
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15664910
|
Honeywell-Fluka
35263-5L-VP |
5 L | VOLPAC™ CONTAINER |
£115.83
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15654910
|
Honeywell-Fluka
35263-5L |
5 L | Plastic Bottle |
£69.69
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15674910
|
Honeywell-Fluka
35263-6X1L |
6 x 1 L | Plastic Bottle |
£120.24
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15624910
|
Honeywell-Fluka
35263-10L-VP |
10 L | VOLPAC™ CONTAINER |
£199.33
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15644910
|
Honeywell-Fluka
35263-4X5L |
4 x 5 L | N/A | ||||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
1310-73-2 | |
NaOH | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
3 mmHg (37°C) |
HNaO | |
0.1M | |
NONH for all modes of transport | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
Sodium hydroxide solution |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide; caustic soda