missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide solution, sodium hydroxide na oh, sodium hydrate, soda, caustic, white caustic, soda lye, aetznatron, caustic soda, ascarite, 1L, CHEBI:32145, Colorless, 39.99g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
£48.40 - £209.00
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | 3548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316190
|
Fisher Chemical
J/7640/15 |
1 L | HDPE plastic bottle |
£48.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10040200
|
Fisher Chemical
J/7640/17 |
2.5 L | HDPE plastic bottle |
£89.50
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10627032
|
Fisher Chemical
J/7640/21 |
5 L | Plastic container |
£161.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10131860
|
Fisher Chemical
J/7640/24 |
10 L | Bag in box | ||||||
Description
HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide solution, sodium hydroxide na oh, sodium hydrate, soda, caustic, white caustic, soda lye, aetznatron, caustic soda, ascarite, 1L, CHEBI:32145, Colorless, 39.99g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
3548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
13 | |
1310-73-2 | |
3548 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
39.99g/mol |
Colorless | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Liquid |