missing translation for 'onlineSavingsMsg'
Learn More
Sodium iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11430021

Sodium iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11430021
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11430021

Brand: Thermo Scientific Alfa Aesar 044766.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of alkyl chlorides into alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300°C to 1304°C
Melting Point 651°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water,alcohol,glycerol and acetone. Insoluble in ether.
Formula Weight 149.89
Odor Odorless
Refractive Index 1.7745
Sensitivity Hygroscopic
Density 3.67
Chemical Name or Material Sodium iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP19L011 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number044766.22,044766.14,044766.36
Lot NumberP30B051 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044766.22,044766.14,044766.36
Lot NumberD17T046 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044766.22,044766.14,044766.36
Lot NumberC24W007 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044766.22,044766.14,044766.36
Lot NumberI06R011 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044766.22,044766.14,044766.36
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium iodide
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.