Learn More
Sodium iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of alkyl chlorides into alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1300°C to 1304°C |
Melting Point | 651°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Soluble in water,alcohol,glycerol and acetone. Insoluble in ether. |
Formula Weight | 149.89 |
Odor | Odorless |
Packaging | Ampuled under Argon |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P19L011 | Certificate of Analysis | 21/03/2025 | 044766.22,044766.14,044766.36 |
P30B051 | Certificate of Analysis | 22/11/2021 | 044766.22,044766.14,044766.36 |
D17T046 | Certificate of Analysis | 22/11/2021 | 044766.22,044766.14,044766.36 |
C24W007 | Certificate of Analysis | 22/11/2021 | 044766.22,044766.14,044766.36 |
I06R011 | Certificate of Analysis | 22/11/2021 | 044766.22,044766.14,044766.36 |
Safety and Handling



- Sodium iodide
- Danger
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : Sodium iodide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.