missing translation for 'onlineSavingsMsg'
Learn More
Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%, Thermo Scientific Chemicals Product Code.: 11396618

Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%, Thermo Scientific Chemicals

Product Code. 11396618
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11396618

Brand: Thermo Scientific Alfa Aesar 012214.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.93
MDL Number MFCD00149170
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdateShow MoreShow Less
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium;dioxido(dioxo)molybdenum;dihydrate
SMILES [Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 100°C-2H2O
Color Green
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5-103.0%
Solubility Information Soluble in water.
Formula Weight 241.95 (205.92 Anhydrous)
Grade ACS
Odor Odorless
Appearance White powder/flake/crystalline/beads
Density 2.71 g/cm3 at 20°C
Chemical Name or Material Sodium molybdenum oxide dihydrate, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN28L028 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number012214.22,012214.36
Lot NumberR16J035 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number012214.22,012214.36
Lot NumberN10J005 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number012214.22,012214.36
Lot NumberZ16G017 Certificate TypeCertificate of Analysis Date15/08/2023 Catalog Number012214.22,012214.36
Lot NumberM18E043 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012214.22,012214.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.