missing translation for 'onlineSavingsMsg'
Learn More
Sodium nitrate, extra pure, Thermo Scientific Chemicals Product Code.: 11904281

Sodium nitrate, extra pure, Thermo Scientific Chemicals

Product Code. 11904281
1 kg, Plastic Bottle
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 11904281

Brand: Thermo Scientific Acros 436090010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1Show MoreShow Less
PubChem CID 24268
ChEBI CHEBI:63005
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point 380.0°C
Melting Point 306.0°C
Color Colorless to White
pH 5.5 to 8.3 (5% aq. soln.)
Physical Form Crystalline Powder and/or Crystals
Quantity 1 kg
Linear Formula NaNO3
Merck Index 15,8778
Solubility Information Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
IUPAC Name sodium nitrate
Formula Weight 84.99
Percent Purity ≥99.0%
Grade Extra Pure
Loss on Drying 2% max. (105°C)
Packaging Plastic Bottle
Chemical Name or Material Sodium nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462905 Certificate TypeCertificate of Analysis Date20/05/2024 Catalog Number436090250,436090010,436090050
Lot NumberA0458290 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number436090250,436090010,436090050
Lot NumberA0458289 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number436090250,436090010,436090050
Lot NumberA0428503 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number436090250,436090010,436090050
Lot NumberA0454616 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number436090250,436090010,436090050
compliance-icons compliance-icons
Product Identifier
  • Sodium nitrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P380-In case of fire: Evacuate area.
  • P375-Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : sodium nitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium nitrate, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.