Learn More
Sodium perchlorate monohydrate, 85+%, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 130.0°C |
Color | White to Yellow |
pH | 6.0 to 8.0 (5% aq. soln. at 25°C) |
Physical Form | Crystalline Powder |
Quantity | 2.5 kg |
Assay Percent Range | 85.0 to 90% (as NaClO4) (Argentometry) |
Molecular Formula | ClNaO4·H2O |
Linear Formula | ClNaO4·H2O |
Merck Index | 15,8784 |
Solubility Information | Solubility in water: 2090g/L (15°C). Other solubilities: soluble in alcohol |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463432 | Certificate of Analysis | 04/06/2024 | 343630025,343631000,343630010 |
A0458081 | Certificate of Analysis | 23/11/2023 | 343630025,343631000,343630010 |
A0450794 | Certificate of Analysis | 02/05/2023 | 343630025,343631000,343630010 |
A0397598 | Certificate of Analysis | 14/12/2022 | 343630025,343631000,343630010 |
A0397597 | Certificate of Analysis | 14/12/2022 | 343630025,343631000,343630010 |
Safety and Handling



- Sodium perchlorate monohydrate
- Danger
- Oxidising solid Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.