missing translation for 'onlineSavingsMsg'
Learn More
Sodium perchlorate monohydrate, 85+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10306691

Sodium perchlorate monohydrate, 85+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10306691
1 kg, Plastic bottle
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10306691

Brand: Thermo Scientific Acros 343630010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-07-3
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrateShow MoreShow Less
PubChem CID 516899
IUPAC Name sodium;perchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]

Specifications

Melting Point 130.0°C
Color White to Yellow
pH 6.0 to 8.0 (5% aq. soln. at 25°C)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 85+%
Molecular Formula ClH2NaO5
Linear Formula ClNaO4·H2O
Solubility Information Solubility in water: 2090g/L (15°C). Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 140.45
Formula Weight 140.46
Percent Purity ≥85%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium perchlorate monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463432 Certificate TypeCertificate of Analysis Date04/06/2024 Catalog Number343630025,343631000,343630010
Lot NumberA0458081 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog Number343630025,343631000,343630010
Lot NumberA0450794 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number343630025,343631000,343630010
Lot NumberA0397598 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number343630025,343631000,343630010
Lot NumberA0397597 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number343630025,343631000,343630010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perchlorate monohydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium perchlorate monohydrate, 85+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.