missing translation for 'onlineSavingsMsg'
Learn More
Sodium perchlorate monohydrate, ACS, 85.0-90.0%, Thermo Scientific Chemicals Product Code.: 15411007

Sodium perchlorate monohydrate, ACS, 85.0-90.0%, Thermo Scientific Chemicals

Product Code. 15411007
1 kg
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15411007

Brand: Thermo Scientific Alfa Aesar 011622.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-07-3
Molecular Formula ClH2NaO5
Molecular Weight (g/mol) 140.45
MDL Number MFCD00149164
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrateShow MoreShow Less
PubChem CID 516899
IUPAC Name sodium;perchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]

Specifications

Melting Point 130°C (decomposition)
Color White
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 85.0-90.0%
Solubility Information Soluble in water.
Formula Weight 140.48 (122.46 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Appearance White crystals
Density 2.02 g/cm3 at 20°C
Chemical Name or Material odium perchlorate monohydrate, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW04K023 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number011622.A1,011622.30
Lot NumberX07K031 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number011622.A1,011622.30
Lot NumberW23K022 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number011622.A1,011622.30
Lot NumberT23K033 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number011622.A1,011622.30
Lot NumberR22K022 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number011622.A1,011622.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium perchlorate monohydrate, ACS, 85.0-90.0%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.