missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, dibasic heptahydrate, 99+%, for analysis, Thermo Scientific Chemicals
£22.21 - £112.56
Chemical Identifiers
CAS | 7782-85-6 |
---|---|
Molecular Formula | H17Na2O11P |
Molecular Weight (g/mol) | 270.08 |
MDL Number | MFCD00149180 |
InChI Key | PYLIXCKOHOHGKQ-UHFFFAOYSA-N |
Synonym | sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate |
PubChem CID | 6096963 |
IUPAC Name | disodium;hydrogen phosphate;heptahydrate |
SMILES | O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10783445
|
Thermo Scientific Acros
206512500 |
250 g | Plastic bottle |
£22.21
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10420101
|
Thermo Scientific Acros
206515000 |
500 g | Plastic bottle |
£31.36
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904291
|
Thermo Scientific Acros
206510025 |
2.5 kg | Plastic bottle |
£112.56
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7782-85-6 | |
270.08 | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
6096963 | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O |
H17Na2O11P | |
MFCD00149180 | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
disodium;hydrogen phosphate;heptahydrate |
Specifications
48.0°C | |
White | |
Crystals | |
H17Na2O11P | |
MFCD00149180 | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
disodium;hydrogen phosphate;heptahydrate | |
6096963 | |
≥99% | |
1.67 |
7782-85-6 | |
9.0 to 9.3 (5% soln. at 20°C) | |
99+% | |
Na2HPO4·7H2O | |
15,8789 | |
Solubility in water: 154g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O | |
270.08 | |
268.07 | |
Analytical | |
Sodium phosphate, dibasic heptahydrate, p.a. |
Safety and Handling
HYGROSCOPIC
missing translation for 'rtecsNumber' : WC4600000
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title