Learn More
Sodium Picosulfate, MedChemExpress
MedChemExpress Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.
Brand: Medchem Express HY-B0544
This item is not returnable.
View return policy
Description
Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.Specifications
Sodium Picosulfate | |
White | |
C18H13NNa2O8S2 | |
Sodium Picosulphate | |
O=S(OC1=CC=C(C=C1)C(C2=CC=CC=N2)C3=CC=C(OS(=O)(O[Na])=O)C=C3)(O[Na])=O | |
481.41 | |
Research | |
Metabolism-protein/nucleotide metabolism |
10040-45-6 | |
Research | |
100 mg | |
DMSO : 100 mg/mL (207.72 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (207.72 mM) | |
481.41 | |
98.0% | |
Solid |