missing translation for 'onlineSavingsMsg'
Learn More

Sodium Picosulfate, MedChemExpress

MedChemExpress Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.

£68.97 - £164.56

Specifications

Chemical Name or Material Sodium Picosulfate
CAS 10040-45-6
Color White
Purity Grade Notes Research
Molecular Formula C18H13NNa2O8S2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16488518
Encompass_Preferred
View Documents
Medchem Express
HY-B0544
10 mg
£68.97
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16418366
Encompass_Preferred
View Documents
Medchem Express
HY-B0544
100 mg
£164.56
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16414397
Encompass_Preferred
View Documents
Medchem Express
HY-B0544
50 mg
£95.59
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.
Specifications

Specifications

Sodium Picosulfate
White
C18H13NNa2O8S2
DMSO : 100 mg/mL (207.72 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (207.72 mM)
481.41
98.0%
Solid
10040-45-6
Research
Sodium Picosulphate
O=S(OC1=CC=C(C=C1)C(C2=CC=CC=N2)C3=CC=C(OS(=O)(O[Na])=O)C=C3)(O[Na])=O
481.41
Research
Metabolism-protein/nucleotide metabolism
Videos
Safety and Handling

Safety and Handling

H302

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.