missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium sulfate decahydrate, extra pure, Thermo Scientific Chemicals
£19.90 - £338.00
Chemical Identifiers
CAS | 7727-73-3 |
---|---|
Molecular Formula | Na2O4S·10H2O |
Molecular Weight (g/mol) | 322.2 |
MDL Number | MFCD00149189 |
InChI Key | RSIJVJUOQBWMIM-UHFFFAOYSA-L |
Synonym | sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o |
PubChem CID | 62649 |
ChEBI | CHEBI:32586 |
IUPAC Name | disodium;sulfate;decahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10760775
|
Thermo Scientific
125012500 |
250 g | Plastic bottle |
£19.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10128043
|
Thermo Scientific Acros
125010010 |
1 kg | Plastic bottle |
£33.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10403415
|
Thermo Scientific Acros
125010025 |
2.5 kg | Plastic bottle |
£57.30
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13498328
|
Thermo Scientific Acros
125010250 |
25 kg | Fibre drum |
£338.00
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7727-73-3 | |
322.2 | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
62649 | |
disodium;sulfate;decahydrate |
Na2O4S·10H2O | |
MFCD00149189 | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o | |
CHEBI:32586 | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Specifications
884.0°C | |
White | |
Crystalline Powder and/or Crystals | |
Na2SO4·10H2O | |
01,1103 | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
disodium;sulfate;decahydrate | |
62649 | |
322.2 | |
Extra Pure |
7727-73-3 | |
5.2 to 9.2 (5% soln.) | |
Na2O4S·10H2O | |
MFCD00149189 | |
14,868 | |
Solubility in water: soluble | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
322.2 | |
CHEBI:32586 | |
≥99.0% | |
Sodium sulfate decahydrate |
RUO – Research Use Only