Learn More
Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals
£20.70 - £117.59
Chemical Identifiers
CAS | 27610-45-3 |
---|---|
Molecular Formula | Na2S |
Molecular Weight (g/mol) | 78.04 |
MDL Number | MFCD00149183 |
InChI Key | GRVFOGOEDUUMBP-UHFFFAOYSA-N |
PubChem CID | 45051681 |
SMILES | [Na+].[Na+].[S--] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10499630
|
Thermo Scientific Acros
133720010 |
1 kg | Plastic bottle |
£20.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10294990
|
Thermo Scientific Acros
133720025 |
2.5 kg | Plastic bottle |
£41.31
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115683
|
Thermo Scientific Acros
133720100 |
10 kg | Fibre drum |
£117.59
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27610-45-3 | |
78.04 | |
GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
[Na+].[Na+].[S--] |
Na2S | |
MFCD00149183 | |
45051681 |
Specifications
27610-45-3 | |
Flakes and Chunks | |
Na2S | |
MFCD00149183 | |
GRVFOGOEDUUMBP-UHFFFAOYSA-N | |
disodium sulfanediide | |
45051681 | |
60 to 64% | |
Sodium sulfide hydrate |
Yellow | |
60 to 64% (Na2S) (Iodimetry) | |
Na2S·xH2O | |
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether | |
[Na+].[Na+].[S--] | |
78.04 | |
78.04 | |
Extra Pure |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Corrosiv
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only