missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, 98.5%, for analysis, anhydrous, Thermo Scientific Chemicals

£19.60 - £172.86

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10422031
View Documents
Thermo Scientific Acros
219270250
25 g Glass Bottle
£19.60
25g
Please to purchase this item. Need a web account? Register with us today!
10320594
View Documents Promotion Details
Thermo Scientific Acros
219272500
250 g Plastic bottle
£20.90
250g
Please to purchase this item. Need a web account? Register with us today!
10032850
View Documents
Thermo Scientific Acros
219270010
1 kg Plastic bottle
£50.55
1kg
Please to purchase this item. Need a web account? Register with us today!
10117743
View Documents
Thermo Scientific Acros
219270050
5 kg Plastic bottle
£172.86
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

7757-83-7
126.04
GEHJYWRUCIMESM-UHFFFAOYSA-L
24437
disodium;sulfite
Na2O3S
MFCD00003503
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
CHEBI:86477
[O-]S(=O)[O-].[Na+].[Na+]

Specifications

>500.0°C
White
98% min. (Iodimetry)
Na2SO3
02,387
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
GEHJYWRUCIMESM-UHFFFAOYSA-L
disodium;sulfite
24437
126.04
Analytical
2.6300g/mL
7757-83-7
Fine Crystals or Crystalline Powder
Na2O3S
MFCD00003503
15, 8811
Solubility in water: 230 g/mL (20°C). Other solubilities: soluble in glycerol, ppractically insoluble in alcohol
[O-]S(=O)[O-].[Na+].[Na+]
126.04
CHEBI:86477
98.5%
2.63
Sodium sulfite, Anhydrous, For analysis
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Contact with acids liberates toxic gas.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 231-821-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.