missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfite, puriss. p.a., Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
4 x 5 kg
50 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Physical Form | Anhydrous |
Quantity | 1 kg |
Linear Formula | Na2SO3 |
UN Number | NONH for all modes of transport |
Formula Weight | 126.04g/mol |
Percent Purity | 98 to 100% (iodometric) |
Grade | puriss. p.a. |
Chemical Name or Material | Sodium Sulfite, Reag. Ph. Eur. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Sulfite, puriss. p.a., Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction