missing translation for 'onlineSavingsMsg'
Learn More
Sodium Sulfite, puriss. p.a., Honeywell Fluka™ Product Code.: 15613450

Sodium Sulfite, puriss. p.a., Honeywell Fluka™

Product Code. 15613450 Shop All Honeywell Fluka Products
5 kg
Click to view available options
Quantity:
500 g
1 kg
5 kg
4 x 5 kg
50 kg
This item is not returnable. View return policy

Product Code. 15613450

Brand: Honeywell-Fluka 314545KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfitShow MoreShow Less
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Anhydrous
Quantity 5 kg
Linear Formula Na2SO3
UN Number NONH for all modes of transport
Formula Weight 126.04g/mol
Percent Purity 98 to 100% (iodometric)
Grade puriss. p.a.
Chemical Name or Material Sodium Sulfite, Reag. Ph. Eur.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Sulfite, puriss. p.a., Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.