Learn More
Sodium tert-butoxide, 97%, Thermo Scientific Chemicals
£44.31 - £486.42
Chemical Identifiers
CAS | 865-48-5 |
---|---|
Molecular Formula | C4H9NaO |
Molecular Weight (g/mol) | 96.105 |
MDL Number | MFCD00040575 |
InChI Key | MFRIHAYPQRLWNB-UHFFFAOYSA-N |
Synonym | sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxideShow More |
PubChem CID | 23676156 |
IUPAC Name | sodium;2-methylpropan-2-olate |
SMILES | CC(C)(C)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448797
|
Thermo Scientific Alfa Aesar
A19942.22 |
100 g |
£44.31
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458797
|
Thermo Scientific Alfa Aesar
A19942.36 |
500 g |
£139.02
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468797
|
Thermo Scientific Alfa Aesar
A19942.0E |
2500 g |
£486.42
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.
Chemical Identifiers
865-48-5 | |
96.105 | |
MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
23676156 | |
CC(C)(C)[O-].[Na+] |
C4H9NaO | |
MFCD00040575 | |
sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxideShow More | |
sodium;2-methylpropan-2-olate |
Specifications
865-48-5 | |
Odorless | |
(CH3)3CONa | |
UN3206 | |
Air and moisture sensitive | |
MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
sodium;2-methylpropan-2-olate | |
23676156 | |
97% |
0.785 | |
C4H9NaO | |
MFCD00040575 | |
3654215 | |
sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide | |
CC(C)(C)[O-].[Na+] | |
96.105 | |
96.11 | |
Sodium tert-butoxide |
Safety and Handling
GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
P210-P235+P410-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314-H500-X
missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 212-741-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Sodium tert-butoxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H228-Flammable solid.
- H251-Self-heating: may catch fire.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium 2-methylpropan-2-olate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.