Learn More
Thermo Scientific Chemicals Sodium tetrathionate dihydrate
£68.24 - £184.00
Chemical Identifiers
CAS | 13721-29-4 |
---|---|
Molecular Formula | H4Na2O8S4 |
Molecular Weight (g/mol) | 306.24 |
MDL Number | MFCD00150868 |
InChI Key | HAEPBEMBOAIUPN-UHFFFAOYSA-L |
Synonym | sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2 |
PubChem CID | 203055 |
IUPAC Name | disodium dihydrate (sulfonatodisulfanyl)sulfonate |
SMILES | O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15783487
|
Thermo Scientific Chemicals
461920250 |
25 g |
£68.24
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15793487
|
Thermo Scientific Chemicals
461921000 |
100 g |
£184.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
13721-29-4 | |
306.24 | |
HAEPBEMBOAIUPN-UHFFFAOYSA-L | |
203055 | |
O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O |
H4Na2O8S4 | |
MFCD00150868 | |
sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2 | |
disodium dihydrate (sulfonatodisulfanyl)sulfonate |
Spécification
13721-29-4 | |
MFCD00150868 | |
Solubility in water: 30.6g/L. | |
O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O | |
306.24 | |
306.27 |
H4Na2O8S4 | |
sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2 | |
HAEPBEMBOAIUPN-UHFFFAOYSA-L | |
disodium dihydrate (sulfonatodisulfanyl)sulfonate | |
203055 | |
Sodium tetrathionate dihydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.