Learn More
Chemical Identifiers
CAS | 7440-23-5 |
---|---|
Molecular Formula | Na |
Molecular Weight (g/mol) | 22.99 |
MDL Number | MFCD00085307 |
InChI Key | KEAYESYHFKHZAL-UHFFFAOYSA-N |
Synonym | natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot |
PubChem CID | 5360545 |
ChEBI | CHEBI:26708 |
IUPAC Name | sodium |
SMILES | [Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10141052
|
Thermo Scientific Acros
370212500 |
250 g | Glass bottle |
£87.23
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914081
|
Thermo Scientific Acros
370210010 |
1 kg | Glass bottle |
£321.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-23-5 | |
22.99 | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
5360545 | |
sodium |
Na | |
MFCD00085307 | |
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot | |
CHEBI:26708 | |
[Na] |
Specifications
Gray or White | |
98.0°C | |
Na | |
MFCD00085307 | |
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot | |
[Na] | |
22.99 | |
CHEBI:26708 | |
≥99.8% | |
Sodium |
7440-23-5 | |
883.0°C | |
Na | |
01,1022; 04,437; 05,589; 07,324; 11,472; 12,126; 16,303 | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
sodium | |
5360545 | |
22.99 | |
Stick |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Sodium, dry stick
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only