Learn More
Sodium thiocyanate, ACS, 98.0% min, Thermo Scientific Chemicals
Description
Sodium thiocyanate is used as a precursor for the synthesis of pharmaceuticals and specialty chemicals. It is used to prepare alkylthiocyanates by reacting with alkyl halides. It is involved nucleophilic substitution reactions with phase-transfer catalysts. It is a common laboratory reagent used to test presence of Fe3+ ions. It acts as a spinning solvent for polyacrylonitrile in fiber industry, photographic stabilizer, corrosion inhibitor, polymer stabilizer and hardening accelerator for cement and concrete mixes. Further, it is also used in both the crystallization and purification of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 287°C |
Color | White |
Physical Form | Crystals and/or Chunks |
Quantity | 500 g |
Assay Percent Range | 98% min. |
Beilstein | 3594965 |
Solubility Information | Soluble in water,acetone,alcohol.Soluble in water,acetone,alcohols and ammonia. |
IUPAC Name | sodium;thiocyanate |
Formula Weight | 81.07 |
Grade | ACS |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q09L025 | Certificate of Analysis | 10/04/2025 | 033388.A3,033388.36,033388.22 |
Y22K031 | Certificate of Analysis | 21/01/2025 | 033388.A3,033388.36,033388.22 |
M16L021 | Certificate of Analysis | 18/01/2025 | 033388.A3,033388.36,033388.22 |
U23K061 | Certificate of Analysis | 16/09/2024 | 033388.A3,033388.36,033388.22 |
T29K044 | Certificate of Analysis | 02/08/2024 | 033388.A3,033388.36,033388.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.