Learn More
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
£27.10 - £917.00
Chemical Identifiers
CAS | 5188-07-8 |
---|---|
Molecular Formula | CH3NaS |
Molecular Weight (g/mol) | 70.09 |
MDL Number | MFCD00174316 |
InChI Key | RMBAVIFYHOYIFM-UHFFFAOYSA-M |
PubChem CID | 4378561 |
SMILES | C[S-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10361044
|
Thermo Scientific Acros
305710010 |
1 g | Glass bottle |
£27.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10043710
|
Thermo Scientific Acros
305710050 |
5 g | Glass bottle |
£79.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10217223
|
Thermo Scientific Acros
305710250 |
25 g | Glass bottle |
£285.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11955741
|
Thermo Scientific Acros
305711000 |
100 g | Glass bottle |
£917.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5188-07-8 | |
70.09 | |
RMBAVIFYHOYIFM-UHFFFAOYSA-M | |
C[S-].[Na+] |
CH3NaS | |
MFCD00174316 | |
4378561 |
Specifications
5188-07-8 | |
Authentic | |
CH3NaS | |
MFCD00174316 | |
Solubility in water: freely soluble. | |
C[S-].[Na+] | |
70.09 | |
70.09 | |
Pure | |
Sodium thiomethoxide |
White | |
94 to 106% (Iodimetry) | |
CH3SNa | |
sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium | |
RMBAVIFYHOYIFM-UHFFFAOYSA-M | |
sodium;methanethiolate | |
4378561 | |
95% | |
Powder |
Safety and Handling
GHS H Statement
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 225-969-9
RUO – Research Use Only