Learn More
Sofosbuvir-d6, MedChemExpress
MedChemExpress Sofosbuvir D6 (PSI-7977 D6) is the deuterium labeled Sofosbuvir. Sofosbuvir (PSI-7977) is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.
Brand: Medchem Express HY-15005S1
This item is not returnable.
View return policy
Description
Sofosbuvir D6 (PSI-7977 D6) is the deuterium labeled Sofosbuvir. Sofosbuvir (PSI-7977) is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.Specifications
Sofosbuvir-d6 | |
5 mg | |
Research | |
PSI-7977-d6 GS-7977-d6 | |
535.49 | |
95.0% | |
Solid |
1868135-06-1 | |
White | |
C22H23D6FN3O9P | |
O=C1N([C@H]2[C@]([C@H](O)[C@@H](CO[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)=O)O2)(C)F)C=CC(N1)=O | |
535.49 | |
Research |