missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir-d6, MedChemExpress

MedChemExpress Sofosbuvir D6 (PSI-7977 D6) is the deuterium labeled Sofosbuvir. Sofosbuvir (PSI-7977) is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.

£480.37 - £1936.00

Specifications

Chemical Name or Material Sofosbuvir-d6
CAS 1868135-06-1
Color White
Purity Grade Notes Research
Molecular Formula C22H23D6FN3O9P
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16458877
Encompass_Preferred
View Documents
Medchem Express
HY-15005S1
5 mg
£1936.00
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16451469
Encompass_Preferred
View Documents
Medchem Express
HY-15005S1
1 mg
£480.37
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sofosbuvir D6 (PSI-7977 D6) is the deuterium labeled Sofosbuvir. Sofosbuvir (PSI-7977) is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.
Specifications

Specifications

Sofosbuvir-d6
White
C22H23D6FN3O9P
O=C1N([C@H]2[C@]([C@H](O)[C@@H](CO[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)=O)O2)(C)F)C=CC(N1)=O
535.49
Research
1868135-06-1
Research
PSI-7977-d6 GS-7977-d6
535.49
95.0%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.