Learn More
Hamilton™ SoftGrip™ Adjustable Volume Multiple Channel Pipette
With a dual indicator that improves the volume adjustment accuracy by removing parallax errors.
Brand:  Hamilton™ 55019-52
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12326239
  • £611.00 / Each
Estimated Shipment: 02-10-2024
to see stock.
Add to basket
Description
Description
  • Wide range of volumes available
  • 8 or 12 channels
  • Unique, long tip ejector lever and movable lower body can be rotated a full 360° relative to handle
  • Isolated volume adjustment so there is no need for a locking mechanism to prevent accidental volume adjustment
  • Universal calibration key that reduces the need for costly calibration services
  • Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
Specifications
0.50±%
Autoclavable
8
Multi-Channel Adjustable Volume Pipette
0.30±%
8 channels
Brick Red
SoftGrip™
30 to 300 μL
1/Pk.