Learn More
Hamilton™ SoftGrip™ Adjustable Volume Multiple Channel Pipette
With a dual indicator that improves the volume adjustment accuracy by removing parallax errors.
Brand: Hamilton™ 55019-54
Code : NEW
Additional Details : Weight : 0.55900kg
Description
- Wide range of volumes available
- 8 or 12 channels
- Unique, long tip ejector lever and movable lower body can be rotated a full 360° relative to handle
- Isolated volume adjustment so there is no need for a locking mechanism to prevent accidental volume adjustment
- Universal calibration key that reduces the need for costly calibration services
- Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
1.00±% | |
Autoclavable | |
12 | |
Multi-Channel Adjustable Volume Pipette | |
0.50±% |
12 channels | |
Sandstone | |
SoftGrip™ | |
5 to 50 μL | |
1/Pk. |