missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Soil Hydrometer

The Soil Hydrometer assesses the suspension's specific gravity or the quantity of soil colloids in soil samples, varying based on the chosen model.

Brand:  VEE GEE 67151H

 View more versions of this product

Product Code. 17836503

  • £53.70 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Approx. Length: 280 mm
  • Calibrated for use at 20°C/68°F
Specifications

Specifications

Soil Hydrometer
0.001
The Soil ASTM Specific Gravity hydrometer measures the specific gravity of soil samples in suspension.
20°C (68°F)
151H
28 cm
0.995 to 1.038
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.