Learn More
3M™ Solus™ Safety Glasses
3M™ Solus™ Safety Glasses are stylish and lightweight for improved comfort and wearer acceptance.
£166.11 - £293.00
Specifications
Certifications/Compliance CE Approved, EN 166: 2001
Product Type Safety glasses
Includes Microfiber storage bag
Lens Material Polycarbonate
View More Specs
Products 7
Product Code Brand Lens Tint Lens Coating Price Quantity & Availability  
Product Code Brand Lens Tint Lens Coating Price Quantity & Availability  
15719677
View Documents
3M™
7000032474
Bronze Anti-fog, Scratch-resistant N/A N/A N/A
15729677
View Documents
3M™
7000032475
Amber Anti-fog, Scratch-resistant N/A N/A N/A
15739677
View Documents
3M™
7000032476
Indoor/Outdoor, Mirror Scratch-resistant N/A N/A N/A
15739807
View Documents
3M™
7000061911
Clear Scratch-resistant N/A N/A N/A
15749677
View Documents
3M™
7000032478
Blue, Mirror Scratch-resistant N/A N/A N/A
15749807
View Documents
3M™
7000061912
Red, Mirror Scratch-resistant N/A N/A N/A
15759807
View Documents
3M™
7000061914
Clear Anti-fog, Scratch-resistant N/A N/A N/A
Description
Description
  • Lens curve has been calculated to optimize the fitting, field of vision and coverage of the eyes
  • Extremely stylish and lightweight glasses
  • Soft inner padding of the temple, leading to increased comfort
  • Supplied with a microfiber storage bag for cleaning and protection
  • Anti-scratch and anti-fog coating for increased durability and improved vision
Specifications
Specifications
CE Approved, EN 166: 2001
Safety glasses
Microfiber storage bag
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Solus™ Safety Glasses