Learn More
asecos™ Sorbents Transport Trolley
Transport trolly for fast access in case of leakage.
Brand:  asecos™ 14411
 View more versions of this product
Product Code. 17179150
  • £579.00 / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • content: 6 cushions, 3 socks, 20 mats, 3 pair protective gloves, 5 plastic bags, 1 pair full vision safety glasses with elastic strap
  • 410 x 590 x 950 (W x D x H)
Specifications
Specifications
Grey
Polyethylene
Oil Sorbents
410.00 mm
410.00 mm
950.00 mm
590.00 mm
35 kg
41 L
Spill Control Cart
16.14 in.
16.14 in.
37.40 in.
23.23 in.
77.16 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
asecos™ Sorbents Transport Trolley > For Use with Oil Sorbents