Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Brand:  Thermo Scientific™ 75007572
Code : 51
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10573992
  • £111.00 / Each
In Stock
to see stock.
Description
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Specifications
15 mL DIN
4-place Swing rotor with round bucket
15 mL