Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
£111.03 - £251.73
Specifications
For Use With (Equipment) 4-place Swing rotor with round bucket
View More Specs
Products 6
Product Code Brand Tube Type Tube Size Price Quantity & Availability  
Product Code Brand Tube Type Tube Size Price Quantity & Availability  
10469502
View Documents Promotion Details
Thermo Scientific™
75007577
Conical 50 mL
£117.82
Each
10573992
View Documents Promotion Details
Thermo Scientific™
75007572
15 mL DIN 15 mL
£111.03
Each
10260452
View Documents Promotion Details
Thermo Scientific™
75007578
Microtube 1.5/2 mL
£251.73
Each
10351192
View Documents Promotion Details
Thermo Scientific™
75007579
7 mL Blood 7 mL
£114.43
Each
10113732
View Documents Promotion Details
Thermo Scientific™
75007621
Conical 15 mL
£117.82
Each
10289871
View Documents Promotion Details
Thermo Scientific™
75007571
7 mL DIN 7 mL
£112.73
Each
Description
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Specifications
4-place Swing rotor with round bucket
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets