missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SOURCE™ Tricorn™ Columns for IEX

Product Code. p-3676618 Shop All Cytiva Products
Click to view available options
Product Type:
Source 15Q prepacked column
Source 15S prepacked column
Ligand Type:
Quaternary Ammonium
Sulphonate group
pH Range:
2 to 12
2 to 13
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10248574

Brand: Cytiva 17518201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Tricorn columns have Valco fittings for AKTAdesign™ systems and are supplied with M6 connectors for FPLC Systems, instruction book, and two stop plugs attached to the column.

Source 15Q 4.6/100 PE is a strong anion exchanger prepacked in a Tricorn column for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.

Pre-packed ion exchange columns for high performance ion exchange chromatography of proteins, peptides, polynucleotides and other biomolecules

  • 4.6/100 PEEK columns provide fast, high-resolution purifications at lab scale and easy scale-up
  • Available with quaternary ammonium strong anion (15Q) or methyl sulfonate strong cation (15S) exhanger
  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics
  • Sample loading up to 25mg of protein/mL
  • High stability in organic solvents and at pH extremes

Recommended Storage

4 to 30°C, 20% Ethanol

TRUSTED_SUSTAINABILITY

Specifications

Buffer All common ion exchange Buffers
Product Type Source 15S prepacked column
Bed Volume 1.7mL
Diameter (English) Inner 0.18 in.
Diameter (Metric) Inner 4.6mm
Quantity 1 x 1.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <5 mL/min.
Material PEEK
Ligand Type Sulphonate group
Max. Pressure 40 bar
pH Range 2 to 13
Particle Size 15μm
Support Type Polystyrene/Divinylbenzene
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) High performance ion exchange chromato graphy of Proteins, Peptides, Polynucleotides and other biomolecules
Show More Show Less
compliance-icons
Product Identifier
  • Source 15S 4.6/100 PE
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.