Learn More
Lenz Laborglasintrumente™ Soxhlet Extractors with Allihn Condenser
For the extraction from solids with solvents
Brand:  Lenz Laborglasintrumente™ 05365049
Product Code. 10346121
  • £322.40 / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The solvent is recirculated continuously and distilled from the extracted sample before
    contacting the solid again
  • The extract is concentrated in the distillation flask
  • The method allows an effective separation with relatively small amounts of extracting solvents
  • The complete apparatuses (all made of DURAN™ tubing) are consisting of:
    • Extractor in compact and robust design, with flask connection NS 29/32
    • Allihn condenser with thread GL 14
    • Round bottom flask
Specifications
Specifications
DURAN Tubing
250 mL extractor, Allihn condenser (NS 45/40), and 500 mL round bottom flask
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente™ Soxhlet Extractors with Allihn Condenser > Includes: 250 mL extractor, Allihn condenser (NS 45/40), and 500 mL round bottom flask