missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Replacement Pen Tips for Electronic Colony Counter

Replacement pen tips fit Scienceware Colony Counter Cat. No. 07910018.

Brand:  Bel-Art™ H37862-0150

Code : 03

Additional Details : Weight : 1.81440kg

 View more versions of this product

Product Code. 12366417

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • The long life, red and black markers tips work on plastic or glass and are oil and water resistant

Specifications

Specifications

Replacement Pen Tip
0.75 in.
1.8 in.
0.75 in.
Accurate sample counting, perfect for Plastic and Glass surfaces
Red
1.9cm
4.6 cm
1.9cm
Scienceware™ electronic colony counter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers