missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Horizontal Models

A UV light-blocking, dry environment

Brand:  Bel-Art™ F42074-0000

 View more versions of this product

Product Code. 11377184

  • £844.47 / Each
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.

  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50%, making them outstanding choices for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • Easy-to-view hygrometer is located in the door front
  • 4.0 horizontal has a wider footprint, lower height and pull down “oven style” door
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626

Dimensions

  • Secador 1.0: 0.74 cu. ft. internal volume, 16.3L x 13.4W x 8.4 in.H
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L x 13.4W x 12.4 in.H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L x 13.4W x 16.4 in.H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L x 13.4W x 20.4 in.H

Order Info

Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100.

Specifications

Specifications

Not Autoclavable
Horizontal
Bottom-hinged and gasketed
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
2
Adjustable Perforated
20.4 in.
51.8 cm
13.4 in.
34.1 cm
41.4 cm
Clear
Copolyester
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
4.0
Chemicals, stains and fine cracks
1.9 cu. ft.
8.9 in.
41.7 cm
16.4 in.
16.3 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.