missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts

Protects electromechanical and optical equipment from exposure to vibration. Bel Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts facilitates optimal equipment use.

Brand:  Bel-Art™ H18386-0001

 View more versions of this product

Product Code. 12330679

  • £1245.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

Protects electromechanical and optical equipment from exposure to vibration. Bel Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts facilitates optimal equipment use.

  • Polished surface resists scratches and chemical attack 6.4cm (2½") thick platforms
  • Molded of dense black & white terrazzo
  • Four Vibro-Absorber™ cushions with neoprene feet support up to 16kg (35 lb)
  • Larger size will absorb vibrations down to 13 Hz with a 16kg (35lb) load; smaller size will absorb vibrations down to 16 Hz. with a 16kg (35lb) load
  • With lighter loads, the minimum frequency of damping for both sizes will increase to a value between 16 Hz. and 25 Hz
  • Shipping weights: large 39kg (87lb), small 19kg (42lb)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.