missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments SP Standard Single-channel Pump Head

Recommended for standard applications

Brand:  Heidolph Instruments 523-43010-00

345.76 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.


 View more versions of this product

Product Code. 10748021

  • £450.00 / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Recommended for standard applications
  • Convex rollers do not squeeze or crush organic cell cultures
  • Pump head features ball bearings
  • Rollers made of stainless steel and polyamide
  • Depending on drive and tubing, the flow rate ranges from 2 to 4151 mL/min.
Specifications

Specifications

2.0 to 4151 mL/min.
Single-channel pumps
Pump Head
Standard applications
1.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers