missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Spill Trays

Spill tray with black base and yellow platform.

£168.00 - £334.00

Specifications

Color Yellow, Black
Material Polyethylene
Type Spill Tray
For Use With (Application) Beverage and Food Processing, Chemical, Healthcare, Manufacturing, Pharmaceutical, Laboratory
View More Specs

Products 4
Product Code Brand Capacity (Metric) Length (Metric) Width (Metric) Weight (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Length (Metric) Width (Metric) Weight (Metric) Price Quantity & Availability  
17801755
View Documents
Brady™
197423
20 L 40 cm 60 cm 3 kg
£168.00
Each
In Stock
to see stock.
Edge
Add to basket
 
17881745
View Documents
Brady™
197579
30 L 40.5 cm 80.5 cm 4.5 kg
£202.00
Each
In Stock
to see stock.
Add to basket
 
17891745
View Documents
Brady™
306709
40 L 80 cm 60 cm 6.5 kg
£246.00
Each
In Stock
to see stock.
Add to basket
 
17821755
View Documents
Brady™
306710
60 L 100 cm 60 cm 9 kg
£334.00
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • For absorbents and spill control
Specifications

Specifications

Yellow, Black
Polyethylene
Spill Tray
Beverage and Food Processing, Chemical, Healthcare, Manufacturing, Pharmaceutical, Laboratory
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ SPC™ Absorbents Spill Trays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.