Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
Brand:  3M™ 7000052909
1019.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15263059
  • £1970.00 / Pack of 180
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Fits a wide range of face shapes and sizes
  • Low breathing resistance filter technology for easier breathing
  • Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
  • An embossed top panel to reduce fogging of eyewear
  • A sculpted nose panel for improved compatibility with eyewear
  • An innovative chin tab for ease of donning and adjustment of the respirator on the face
  • Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
  • Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10
Red
Cup/Molded
Particulate Respirator with Nuisance Organic Vapor Relief
Yes
Electret (polyester, polypropylene, and carbon)
Yes
Acid Gas
Cup
No
EN149:2001 + A1:2009
White
Single-use
Standard
FFP3
Cool-flow Valve, Red Color Coded Straps
9936
180/Pk.
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Aura™ Specialty Respirator, Series 9300 > Model: 9936; Valved; For use with: 3M eyewear and hearing protection