missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Specific Gravity Hydrometer, Wide-Range

The Wide-Range Specific Gravity Hydrometer is ideal when you aren't sure of the approximate value of your liquid.

Brand:  VEE GEE 6604-3

 View more versions of this product

Product Code. 17816463

  • £29.95 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Subdivisions: 0.005
  • Approx. Length: 165 mm
  • Calibrated for use at 15.5°C/60°F
Specifications

Specifications

Specific Gravity Hydrometer
16.5 cm
15.6°C (60°F)
0.005
1.400 to 1.650
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VEE GEE Specific Gravity Hydrometer, Wide-Range > Specific Gravity Range: 1.400 to 1.650

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.