missing translation for 'onlineSavingsMsg'
Learn More

Spectrum Labs™ Spectra/Mesh™ Woven Nylon Filter Sheet

Hydrophilic nylon filters with high strength for use in laboratory, industrial, or pharmaceutical applications.

Brand:  Spectrum Labs™ 146519

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15330922

  • £343.00 / Pack of 3
Estimated Shipment: 10-09-2024
to see stock.
Add to basket

Description

Description

  • Hydrophilic
  • Good resistance against corrosives
  • Resistant against pH 3 to 10
  • Sterilize via irradiation
Specifications

Specifications

Nylon
Laboratory, Industrial, Pharmaceutical
100 μm
5 μm
2%
30 x 30 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers