Learn More
Spectrum Labs™ Spectra/Por™ 1 Membrane Disc
Regenerated cellulose membrance for use with Spectrum Equilibrium Dializer and Spectra/Por MacroDializer systems
Brand: Spectrum Labs™ 132476
This item is not returnable.
View return policy
Description
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Specifications
Membrane Disc | |
Regenerated Cellulose | |
For Basic Dialysis |
6 to 8 kDa | |
47 mm |