missing translation for 'onlineSavingsMsg'
Learn More

Spectrum Labs™ Spectra/Por™ Cellulose Regenerated Tubing Membrane

For molecular separation, protein binding studies, electrolyte removal and desalting

Brand:  Spectrum Labs™ 132584

 View more versions of this product

Product Code. 11485879

  • £434.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Pure regenerated natural cellulose membranes with 1kDa to 50kDa MWCO. Transparent and flexible except Spectra/Por™ 5. Low molecular weight compounds in aqueous solutions diffuse through the membrane, higher molecular weights, such as proteins and bacteria, are retained by the membrane. Convenient dispenser boxes protect unused membranes. Glycerol (where used) preserves membrane and must be removed before use. Soak membrane for three hours in deionised water, then rinse well with running water. Spectra/Por™ 5 is reinforced with a layer of porous paper, to give a thick, rigid membrane for use in high shear environments.
  • Spectra/Por™ 1, 2, 3, 4, 5, supplied dry with glycerol
  • Spectra/Por™ 6, pre-wetted supplied in 0.1% sodium azide
  • Spectra/Por™ 7, pre-treated to reduce trace contaminants supplied in 0.1% sodium azide
For molecular separation, protein binding studies, electrolyte removal and desalting.
Specifications

Specifications

Regenerated Tubing Membrane
Cellulose
10 m
Transparent
1 Each
8 kDa
25.5 mm
5.1 mL/cm
60°C
For molecular separation, protein binding studies, electrolyte removal and desalting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions