missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Spectra/Por™ Cellulose Regenerated Tubing Membrane
For molecular separation, protein binding studies, electrolyte removal and desalting
£323.00 - £434.00
Specifications
Product Type | Regenerated Tubing Membrane |
---|---|
Material | Cellulose |
Color | Transparent |
Temperature Range (Metric) | 60°C |
Quantity | 1 Each |
Product Code | Brand | Capacity (Metric) | MWCO | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | MWCO | Length (Metric) | Price | Quantity & Availability | |||
11415909
|
Spectrum Labs™
132539 |
0.32 mL/cm | 50 kDa | 5 m | N/A | ||||
11485889
|
Spectrum Labs™
128158 |
1.1 mL/cm | 15 kDa | 10 m | N/A | ||||
11465879
|
Spectrum Labs™
128058 |
1.1 mL/cm | 8 kDa | 10 m |
£382.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11465899
|
Spectrum Labs™
128218 |
1.1 mL/cm | 25 kDa | 10 m |
£434.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11445919
|
Spectrum Labs™
128358 |
1.1 mL/cm | 8 kDa | 5 m |
£323.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11435879
|
Spectrum Labs™
132579 |
0.45 mL/cm | 8 kDa | 10 m |
£382.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11495859
|
Spectrum Labs™
132757 |
46 mL/cm | 12 to 14 kDa | 15 m |
£338.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11415899
|
Spectrum Labs™
132566 |
6.4 mL/cm | 15 kDa | 10 m |
£382.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11485879
|
Spectrum Labs™
132584 |
5.1 mL/cm | 8 kDa | 10 m |
£434.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Pure regenerated natural cellulose membranes with 1kDa to 50kDa MWCO. Transparent and flexible except Spectra/Por™ 5. Low molecular weight compounds in aqueous solutions diffuse through the membrane, higher molecular weights, such as proteins and bacteria, are retained by the membrane. Convenient dispenser boxes protect unused membranes. Glycerol (where used) preserves membrane and must be removed before use. Soak membrane for three hours in deionised water, then rinse well with running water. Spectra/Por™ 5 is reinforced with a layer of porous paper, to give a thick, rigid membrane for use in high shear environments.- Spectra/Por™ 1, 2, 3, 4, 5, supplied dry with glycerol
- Spectra/Por™ 6, pre-wetted supplied in 0.1% sodium azide
- Spectra/Por™ 7, pre-treated to reduce trace contaminants supplied in 0.1% sodium azide
Specifications
Regenerated Tubing Membrane | |
Transparent | |
1 Each |
Cellulose | |
60°C | |
For molecular separation, protein binding studies, electrolyte removal and desalting |