missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra Mesh™ Woven, Nylon Filters

Consist of individual strands woven into a mesh screen

Brand:  Spectrum™ 148132

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15300952

  • £144.00 / Pack of 10
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

Combines high tensile strength with flexibility

  • Water Affinity: Hydrophillic
  • Strength: High
  • Adsorption: High
  • Best Resistance: Corrosives
  • pH Resistance: pH 3-10
  • Thermal Stability: up to 180°C
  • Sterilization: Irradiation
Specifications

Specifications

Disc
10 μm
45 μm
Nylon
Hazardous waste, drinking water purification, Acid rain measurement, suspended soils, air/asbestos, coal dust analysis, oceanography, trace element analysis
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers