missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra Mesh™ Woven, Nylon Filters
Consist of individual strands woven into a mesh screen
Brand: Spectrum™ 145 809
Code : NEW
Additional Details : Weight : 0.02970kg
Description
Combines high tensile strength with flexibility
- Water Affinity: Hydrophillic
- Strength: High
- Adsorption: High
- Best Resistance: Corrosives
- pH Resistance: pH 3-10
- Thermal Stability: up to 180°C
- Sterilization: Irradiation
Specifications
Disc | |
30 μm | |
64 μm |
Nylon | |
Hazardous waste, drinking water purification, Acid rain measurement, suspended soils, air/asbestos, coal dust analysis, oceanography, trace element analysis | |
55 mm |