missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Standard RC Ready-to-use Spectra/Por™ Dialysis Sacks

For large sample volumes

Brand:  Spectrum™ 132 707

 View more versions of this product

Product Code. 15310772

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Material: Regenerated Cellulose
  • Length: 60cm (23.62 in.)
  • Sealed at one end, open at the other
  • Funnel attached to top for easy sample filling
  • Supplied in sodium azide 0.05% (must be rinsed prior to use)

Specifications

Specifications

Dialysis Sacks
Spectra/Por 4
29 mm
23.6 in.
45 mm
0.05% sodium azide preservant
For large sample volumes, funnel attached to top for easy sample filling
12 to 14 kDa
Regenerated Cellulose
1.14 in.
600 mm
50 to 400 mL
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers